KEY358812835 Key Organics / BIONET

ethyl (2E)-3-[1-methyl-4-(4-methylbenzoyl)-1H-pyrrol-2-yl]prop-2-enoate

  • MDL number MFCD21333202
  • Cas Number : 428872-08-6
  • Molecular Weight : 297.354

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94 - 96
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94785

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0203_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart