KEY358812840 Key Organics / BIONET

3-(trifluoromethyl)quinoxaline-2-carboxylic acid

  • MDL number MFCD11869266
  • Cas Number : 1142190-60-0
  • Molecular Weight : 242.157

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164 (dec)
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94797

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0705_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart