KEY358812844 Key Organics / BIONET

7H-pyrrolo[2,3-d]pyrimidine-4-carboxylic acid

  • MDL number MFCD13193623
  • Cas Number : 1005206-17-6
  • Molecular Weight : 163.136

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 (dec)
Purity 97
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94801

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AG-0710_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart