KEY358812845 Key Organics / BIONET

1-(2,6-dichloropyridin-3-yl)ethan-1-one

  • MDL number MFCD11847148
  • Cas Number : 412018-50-9
  • Molecular Weight : 190.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 32 - 34
Iupac Name 1-(2,6-dichloro-3-pyridinyl)ethanone
Inchi Code 1S/C7H5Cl2NO/c1-4(11)5-2-3-6(8)10-7(5)9/h2-3H,1H3
Inchi Key XOEFQKGFPDZVKF-UHFFFAOYSA-N

Description

Melting Point 32 - 34
Purity 97%
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94802

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AG-0712_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart