KEY358812846 Key Organics / BIONET

2,6-bis(trifluoromethyl)pyridine-4-carboxylic acid

  • MDL number MFCD11501683
  • Cas Number : 1092343-70-8
  • Molecular Weight : 259.107

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 125
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7403

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0856_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart