KEY358812849 Key Organics / BIONET

methyl 2-bromo-4-fluoro-5-methoxybenzoate

  • MDL number MFCD20482372
  • Cas Number : 1007455-22-2
  • Molecular Weight : 263.062

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36 - 38
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101907

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LF-0032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart