KEY362932565 Key Organics / BIONET

[(3-methoxy-4-methylphenyl)methyl](propyl)amine

  • MDL number MFCD11212463
  • Cas Number : 1094641-71-0
  • Molecular Weight : 193.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94780

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0073_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart