KEY362932566 Key Organics / BIONET

2-bromo-5-(3-methoxyphenoxy)-1,3,4-thiadiazole

  • MDL number MFCD20368314
  • Cas Number : 1344306-45-1
  • Molecular Weight : 287.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72 - 73
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94781

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0079_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart