KEY362932568 Key Organics / BIONET

ethyl 6-chloro-2-methylpyridine-3-carboxylate

  • MDL number MFCD11616141
  • Cas Number : 31163-12-9
  • Molecular Weight : 199.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40 - 42
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94788

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0209_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart