KEY362932572 Key Organics / BIONET

(2Z)-3-[(4-chloro-3-nitrophenyl)carbamoyl]prop-2-enoic acid

  • MDL number MFCD00566910
  • Cas Number : 1383931-94-9
  • Molecular Weight : 270.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162 - 166
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94804

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart