KEY362932574 Key Organics / BIONET

ethyl 3-amino-1-methyl-6-(trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-2-carboxylate

  • MDL number MFCD21605748
  • Cas Number : 1370587-21-5
  • Molecular Weight : 287.242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106 - 108
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94807

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart