KEY362932584 Key Organics / BIONET

tert-butyl 4-[(2-aminopyridin-3-yl)amino]piperidine-1-carboxylate

  • MDL number MFCD13184780
  • Cas Number : 781649-86-3
  • Molecular Weight : 292.383

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142 - 146
Purity 97
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95238

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BG-0046_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart