KEY362932587 Key Organics / BIONET

tert-butyl 4-{2-oxo-1H,2H,3H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate

  • MDL number MFCD13184781
  • Cas Number : 781649-87-4
  • Molecular Weight : 318.377

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201 - 204
Purity 97
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95245

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BG-0067_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart