KEY362932588 Key Organics / BIONET

imidazo[1,2-a]pyridine-7-carbonitrile

  • MDL number MFCD09994633
  • Cas Number : 952566-04-0
  • Molecular Weight : 143.149

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133 - 134
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95253

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BG-0205_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart