KEY362932600 Key Organics / BIONET

1-(4-acetyl-5-nitro-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one

  • MDL number MFCD21605753
  • Cas Number : 1269822-93-6
  • Molecular Weight : 299.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 127
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102007

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LF-0721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart