KEY366276599 Key Organics / BIONET

CIS-4-azatricyclo[5.2.2.0^{2,6}]undecane-3,5- dione

  • MDL number MFCD22126132
  • Cas Number : 85922-86-7
  • Molecular Weight : 179.219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108650

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4373_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart