KEY366276600 Key Organics / BIONET

methyl 4-[(6-chloropyridin-3-yl)methyl]benzoate

  • MDL number MFCD22126099
  • Cas Number : 1370587-28-2
  • Molecular Weight : 261.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 83
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94808

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart