KEY366276604 Key Organics / BIONET

ethyl 2-chloro-5-nitropyridine-4-carboxylate

  • MDL number MFCD22126101
  • Cas Number : 907545-64-6
  • Molecular Weight : 230.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40 - 41
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95233

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BG-0018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart