KEY366276608 Key Organics / BIONET

1-(piperidin-4-yl)-1H,2H,3H-imidazo[4,5-b]pyridin-2-one dihydrochloride

  • MDL number MFCD14705173
  • Cas Number : 781649-84-1
  • Molecular Weight : 291.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 350
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95247

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BG-0075_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart