KEY366276617 Key Organics / BIONET

ethyl 5-methylpyridine-2-carboxylate

  • MDL number MFCD10566273
  • Cas Number : 55876-82-9
  • Molecular Weight : 165.192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Bp: 90 - 92 @ 0.1mmHg
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95272

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BG-0720_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart