KEY366276623 Key Organics / BIONET

methyl 6-bromo-1-benzofuran-2-carboxylate

  • MDL number MFCD22126097
  • Cas Number : 425675-94-1
  • Molecular Weight : 255.067

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107 - 109
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3708

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Z-0931_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart