KEY366276627 Key Organics / BIONET

N-[4-chloro-2-(trifluoromethoxy)phenyl]acetamide

  • MDL number MFCD00276972
  • Cas Number : 175278-36-1
  • Molecular Weight : 253.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128 - 130
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77431

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Y-0806_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart