KEY366276639 Key Organics / BIONET

5-methylpyridine-2-carboxylic acid

  • MDL number MFCD01704358
  • Cas Number : 4434-13-3
  • Molecular Weight : 137.138

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 300
Purity 97
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95884

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/CG-0700_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart