KEY366276640 Key Organics / BIONET

(5-methylpyridin-2-yl)methanol

  • MDL number MFCD09999992
  • Cas Number : 22940-71-2
  • Molecular Weight : 123.155

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Bp: 80 - 82 @ 0.5mmHg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95885

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CG-0704_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart