KEY366276682 Key Organics / BIONET

ethyl 2-(5-methyl-4H-1,2,4-triazol-3-yl)acetate

  • MDL number MFCD11973618
  • Cas Number : 100187-10-8
  • Molecular Weight : 169.184

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108375

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3566_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart