KEY366276685 Key Organics / BIONET

(2S)-4-(4-ethoxyphenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid

  • MDL number MFCD16890155
  • Cas Number : 1252046-18-6
  • Molecular Weight : 445.515

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108570

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4165_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart