KEY366276689 Key Organics / BIONET

(2S,4R)-1-[(tert-butoxy)carbonyl]-4-methylpyrrolidine-2-carboxylic acid

  • MDL number MFCD13185988
  • Cas Number : 364750-80-1
  • Molecular Weight : 229.276

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108644

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4367_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart