KEY366276696 Key Organics / BIONET

{[2-hydroxy-3-(phenylsulfanyl)propyl]sulfanyl}benzene

  • MDL number MFCD22126133
  • Cas Number : 22910-07-2
  • Molecular Weight : 276.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108651

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4374_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart