KEY366276697 Key Organics / BIONET

ethyl (2E)-3-(2-fluoro-5-nitrophenyl)prop-2-enoate

  • MDL number MFCD22124782
  • Cas Number : 91319-50-5
  • Molecular Weight : 239.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108652

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4375_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart