KEY366276700 Key Organics / BIONET

3-(3-fluoro-4-methylphenyl)-1,2,4-oxadiazole

  • MDL number MFCD13181200
  • Cas Number : 1146699-63-9
  • Molecular Weight : 178.166

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108656

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4379_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart