KEY369260624 Key Organics / BIONET

2,3,5-trifluoro-6-hydrazinylpyridin-4-amine

  • MDL number MFCD01862074
  • Cas Number : 105252-94-6
  • Molecular Weight : 178.118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191 - 193
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94859

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-0072_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart