KEY369260632 Key Organics / BIONET

4-{2-methyl-1H-imidazo[4,5-b]pyridin-1-yl}piperidine dihydrochloride

  • MDL number MFCD22373666
  • Cas Number : 1379527-04-4
  • Molecular Weight : 289.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 190 - 195
Iupac Name 2-methyl-1-(4-piperidinyl)-1H-imidazo[4,5-b]pyridine dihydrochloride
Inchi Code 1S/C12H16N4.2ClH/c1-9-15-12-11(3-2-6-14-12)16(9)10-4-7-13-8-5-10;;/h2-3,6,10,13H,4-5,7-8H2,1H3;2*1H
Inchi Key LKYCKGIWRDIZIY-UHFFFAOYSA-N

Description

Melting Point 190 - 195
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96215

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DG-0024_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart