KEY369260636 Key Organics / BIONET

1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD01248822
  • Cas Number : 5932-32-1
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >250
Iupac Name 1,4,5,6-tetrahydrocyclopenta[c]pyrazole-3-carboxylic acid
Inchi Code 1S/C7H8N2O2/c10-7(11)6-4-2-1-3-5(4)8-9-6/h1-3H2,(H,8,9)(H,10,11)
Inchi Key FCYBBDFUBSEGMX-UHFFFAOYSA-N

Description

Melting Point >250
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96226

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DG-0059_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart