KEY369260650 Key Organics / BIONET

1-[(4-chlorophenyl)methyl]-5,5-dimethylimidazolidin-2-one

  • MDL number MFCD22373658
  • Cas Number : 1379527-02-2
  • Molecular Weight : 238.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122 - 126
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96550

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EG-0014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart