KEY369260662 Key Organics / BIONET

1H-pyrrolo[2,3-c]pyridine-7-carboxylic acid

  • MDL number MFCD12924374
  • Cas Number : 945840-82-4
  • Molecular Weight : 162.148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96585

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EG-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart