KEY369260666 Key Organics / BIONET

ethyl 2-methylpyridine-4-carboxylate

  • MDL number MFCD09746244
  • Cas Number : 25635-17-0
  • Molecular Weight : 165.192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96590

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EG-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart