KEY369260669 Key Organics / BIONET

ethyl 5,7-bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylate

  • MDL number MFCD22373660
  • Cas Number : 1379526-96-1
  • Molecular Weight : 338.209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96593

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EG-0715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart