KEY369260671 Key Organics / BIONET

5,7-bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylic acid

  • MDL number MFCD22373654
  • Cas Number : 1379526-94-9
  • Molecular Weight : 310.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171 (dec)
Purity 0.97
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96595

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/EG-0718_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart