KEY369260675 Key Organics / BIONET

2-methylpyridine-4-carbaldehyde

  • MDL number MFCD06410683
  • Cas Number : 63875-01-4
  • Molecular Weight : 121.139

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96600

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EG-0724_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart