KEY370561039 Key Organics / BIONET

2-amino-6-phenylpyrimidin-4-ol

  • MDL number MFCD00296928
  • Cas Number : 56741-94-7
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-6-phenyl-4(3H)-pyrimidinone
Inchi Code 1S/C10H9N3O/c11-10-12-8(6-9(14)13-10)7-4-2-1-3-5-7/h1-6H,(H3,11,12,13,14)
Inchi Key KXYGHDWFGFZJPJ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73460

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0631_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart