KEY370561040 Key Organics / BIONET

2-{[(tert-butoxy)carbonyl]amino}-3,5-dimethylbenzoic acid

  • MDL number MFCD02682184
  • Cas Number : 669713-57-9
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[(tert-butoxycarbonyl)amino]-3,5-dimethylbenzoic acid
Inchi Code 1S/C14H19NO4/c1-8-6-9(2)11(10(7-8)12(16)17)15-13(18)19-14(3,4)5/h6-7H,1-5H3,(H,15,18)(H,16,17)
Inchi Key ITWQZQYNZFDETR-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84138

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6W-0019_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart