KEY370561042 Key Organics / BIONET

4-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]pyrimidin-2-amine

  • MDL number MFCD02090064
  • Cas Number : 308088-19-9
  • Molecular Weight : 302.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]-2-pyrimidinamine
Inchi Code 1S/C14H11ClN4S/c1-8-12(11-6-7-17-14(16)19-11)20-13(18-8)9-2-4-10(15)5-3-9/h2-7H,1H3,(H2,16,17,19)
Inchi Key AEVGCUSOXYKFKL-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93403

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9P-706_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart