KEY370561110 Key Organics / BIONET

4,4,5,5-tetramethyl-2-(propan-2-yl)-1,3,2-dioxaborolane

  • MDL number MFCD05663856
  • Cas Number : 76347-13-2
  • Molecular Weight : 170.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95378

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-2076_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart