KEY370561117 Key Organics / BIONET

3-bromo-2-ethoxypyridine

  • MDL number MFCD00234309
  • Cas Number : 57883-25-7
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store under N2
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95385

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-2085_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart