KEY370561157 Key Organics / BIONET

N-[3-(dimethylamino)propyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD05864306
  • Cas Number : 832114-10-0
  • Molecular Weight : 332.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 99
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95425

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-2130_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart