KEY371462891 Key Organics / BIONET

methyl 2-(3-formylphenoxy)acetate

  • MDL number MFCD04227169
  • Cas Number : 37748-10-0
  • Molecular Weight : 194.186

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93296

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9P-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart