KEY371462893 Key Organics / BIONET

8-chloro-6-(trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyridin-2-amine

  • MDL number MFCD22383935
  • Cas Number : 1206640-61-0
  • Molecular Weight : 236.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94771

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AG-0031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart