KEY371462898 Key Organics / BIONET

2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-1,3-benzodiazole

  • MDL number MFCD22383952
  • Cas Number : 1393845-65-2
  • Molecular Weight : 297.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97046

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FG-0027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart