KEY371462902 Key Organics / BIONET

4-bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD11046241
  • Cas Number : 943323-92-0
  • Molecular Weight : 231.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227 - 228
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97050

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FG-0213_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart