KEY371462914 Key Organics / BIONET

4-bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD09880123
  • Cas Number : 1000340-34-0
  • Molecular Weight : 322.931

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222 - 224
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97567

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GG-0205_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart